Skip to Content
Merck
All Photos(2)

Key Documents

242063

Sigma-Aldrich

(S)-(−)-N,N-Dimethyl-1-phenylethylamine

97%

Synonym(s):

(S)-(−)-N,N,α-Trimethylbenzylamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€122.00

€122.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€122.00

About This Item

Linear Formula:
C6H5CH(CH3)N(CH3)2
CAS Number:
Molecular Weight:
149.23
Beilstein:
3196218
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

€122.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

liquid

optical activity

[α]20/D −49.2°, c = 1 in methanol

refractive index

n20/D 1.503 (lit.)

bp

81 °C/16 mmHg (lit.)

density

0.899 g/mL at 25 °C (lit.)

functional group

amine
phenyl

storage temp.

2-8°C

SMILES string

C[C@H](N(C)C)c1ccccc1

InChI

1S/C10H15N/c1-9(11(2)3)10-7-5-4-6-8-10/h4-9H,1-3H3/t9-/m0/s1

InChI key

BVURNMLGDQYNAF-VIFPVBQESA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service