Skip to Content
Merck
All Photos(1)

Key Documents

241857

Sigma-Aldrich

Lanthanum hexaboride

powder, 10 μm, 95%

Synonym(s):

Lanthanum boride

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€128.00

€128.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€128.00

About This Item

Linear Formula:
LaB6
CAS Number:
Molecular Weight:
203.77
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

€128.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

Assay

95%

form

powder

reaction suitability

reagent type: catalyst
core: lanthanum

particle size

10 μm

mp

2210 °C

density

2.61 g/cm3 at 25 °C (lit.)

SMILES string

[La++].B12B3[B-]14B5B4[B-]235

InChI

1S/B6.La/c1-2-5(1)3-4(5)6(1,2)3;/q-2;+2

InChI key

VJXOKIULCJPJLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

One of the best thermionic emitters. Effective in improving diamond nucleation in the chemical vapor deposition of diamond on substrate surfaces.[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maeda, H. et al.
Journal of Crystal Growth, 121, 507-507 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service