241466
Chromium(III) chloride
35-45 wt. % in H2O
Synonym(s):
Chromic chloride, Chromium trichloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CrCl3
CAS Number:
Molecular Weight:
158.36
MDL number:
UNSPSC Code:
12352302
concentration
35-45 wt. % in H2O
density
2.87 g/mL at 25 °C (lit.)
SMILES string
Cl[Cr](Cl)Cl
InChI
1S/3ClH.Cr/h3*1H;/q;;;+3/p-3
InChI key
QSWDMMVNRMROPK-UHFFFAOYSA-K
Looking for similar products? Visit Product Comparison Guide
Application
The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties.2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service