Skip to Content
Merck
All Photos(1)

Key Documents

241016

Sigma-Aldrich

Methylamine hydrochloride

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NH2 · HCl
CAS Number:
Molecular Weight:
67.52
Beilstein:
3588822
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥99%

bp

225-230 °C/15 mmHg (lit.)

mp

231-233 °C (lit.)

solubility

absolute ethanol: soluble(lit.)
acetone: insoluble(lit.)
chloroform: insoluble(lit.)
diethyl ether: insoluble(lit.)
ethyl acetate: insoluble(lit.)
water: soluble(lit.)

SMILES string

Cl[H].CN

InChI

1S/CH5N.ClH/c1-2;/h2H2,1H3;1H

InChI key

NQMRYBIKMRVZLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methylamine hydrochloride causes the chroloplast to shrink.

Application

Methylamine hydrochloride was used in the synthesis of mephtetramine via Mannich reaction.

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

408.7 °F - closed cup

Flash Point(C)

209.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Izawa et al.
Plant physiology, 41(3), 533-543 (1966-03-01)
Whole chloroplasts isolated from the leaves of spinach (Spinacia oleracea L.) exhibit 2 types of conformational change during electron transport. Amine-uncoupled chloroplasts swell and atebrin-uncoupled chloroplasts shrink. Chloroplasts uncoupled by carbonylcyanide phenylhydrazones and by treatment with ethylenediamine tetraacetic acid do
John D Power et al.
Drug testing and analysis, 6(7-8), 668-675 (2014-02-28)
Three legal highs; nitracaine (3-(diethylamino)-2,2-dimethylpropyl 4-nitrobenzoate), methoxypiperamide (MEOP, (4-methoxyphenyl)(4-methylpiperazin-1-yl)methanone) and mephtetramine (MTTA, 2-((methylamino)methyl)-3,4-dihydronaphthalen-1(2H)-one) appeared in 2013 as new psychoactive substances (NPS) on Internet websites selling 'research chemicals'. These compounds were synthesized and analyzed via our synthesize, analyze, and metabolize (SAM)
Jing-Jing Fang et al.
Waste management (New York, N.Y.), 32(7), 1401-1410 (2012-04-07)
This study investigated the odor compounds from different areas in a landfill site, which included the municipal solid waste (MSW)-related area, the leachate-related area and the sludge-related area. Nine sampling points were placed and 35 types of odorous substances were
Nicolas Fleury-Brégeot et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(31), 9564-9570 (2012-07-07)
Ammoniomethyl trifluoroborates are very powerful reagents that can be used to access biologically relevant aryl- and heteroaryl-methylamine motifs via Suzuki-Miyaura cross-couplings. Until now, this method was limited to the production of tertiary and primary amines. The synthesis of a large
Thomas S Hofer et al.
Molecular bioSystems, 8(11), 2891-2900 (2012-08-01)
Molecular dynamics simulations have been performed to investigate the binding of tris(hydroxymethyl)-aminomethane to the surface of the core domain of the mouse cellular tumor antigen p53 employing the GROMOS and 53A6 force field parameter sets. A close investigation of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service