Skip to Content
Merck
All Photos(1)

Key Documents

240966

Sigma-Aldrich

1-Butanethiol

≥99%

Synonym(s):

Butyl mercaptan, Mercaptan C4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3SH
CAS Number:
Molecular Weight:
90.19
Beilstein:
1730908
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3.1 (vs air)

vapor pressure

83 mmHg ( 37.7 °C)

Assay

≥99%

refractive index

n20/D 1.443 (lit.)

bp

98 °C (lit.)

mp

−116 °C (lit.)

density

0.842 g/mL at 25 °C (lit.)

SMILES string

CCCCS

InChI

1S/C4H10S/c1-2-3-4-5/h5H,2-4H2,1H3

InChI key

WQAQPCDUOCURKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Mather-Mihaich et al.
Comparative biochemistry and physiology. C, Comparative pharmacology and toxicology, 85(2), 427-432 (1986-01-01)
Indicators of free-radical or oxidant-mediated responses were quantified in channel catfish, Ictalurus punctatus, exposed to the organophosphorus herbicide DEF and its metabolite, n-butyl mercaptan (nBM). The activities of the antioxidant enzymes superoxide dismutase (SOD), glutathione peroxidase (GPX) and catalase did
Late acute, delayed neurotoxic and cholinergic effects of S,S,S-tributyl phosphorotrithioite (Merphos) in hens.
M B Abou-Donia et al.
Toxicology and applied pharmacology, 53(3), 439-457 (1980-05-01)
Y Cao et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 57(1), 27-34 (2001-02-24)
The oxidative stability of self-assembled monolayer (SAM) of thiols on silver was examined with matrix-assisted laser desorption ionization (MALDI) time-of-flight (TOF) mass spectrometry (MS). A benzenesulfinate (BS) monolayer on silver was also prepared and investigated with MALDI-TOF-MS and surface-enhanced Raman
Rodolphe Obeid et al.
Journal of colloid and interface science, 340(2), 142-152 (2009-10-03)
We describe herein the properties at the air/water (A/W) interface of hydrophobically end-modified (HM) poly(2-isopropyl-2-oxazoline)s (PiPrOx) bearing an n-octadecyl chain on both termini (telechelic HM-PiPrOx) or on one chain end (semitelechelic HM-PiPrOx) for different subphase temperatures and spreading solvents using
Nicolò Dossi et al.
Lab on a chip, 12(1), 153-158 (2011-11-15)
A sensitive and fast-responding membrane-free amperometric gas sensor is described, consisting of a small filter paper foil soaked with a room temperature ionic liquid (RTIL), upon which three electrodes are screen printed with carbon ink, using a suitable mask. It

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service