Skip to Content
Merck
All Photos(1)

Key Documents

240125

Sigma-Aldrich

Hydroquinone

≥99%

Synonym(s):

1,4-Benzenediol, 1,4-Dihydroxybenzene, HQ

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,4-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein:
605970
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor density

3.81 (vs air)

vapor pressure

1 mmHg ( 132 °C)

Assay

≥99%

autoignition temp.

930 °F

bp

285 °C (lit.)

mp

172-175 °C (lit.)

SMILES string

Oc1ccc(O)cc1

InChI

1S/C6H6O2/c7-5-1-2-6(8)4-3-5/h1-4,7-8H

InChI key

QIGBRXMKCJKVMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

329.0 °F - closed cup

Flash Point(C)

165 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quan Cheng Chen et al.
Journal of natural products, 71(6), 995-999 (2008-05-29)
Five new pyronane-type monocyclic monoterpenoids, jasminodiol (1), jasminoside H (6), 6'-O-sinapoyljasminoside A (7), 6'-O-sinapoyljasminoside C (8), and jasminoside I (9), together with four known analogues, were isolated from the fruit of Gardenia jasminoides. The structures of the new metabolites were
Woo Duck Seo et al.
European journal of medicinal chemistry, 45(5), 2010-2017 (2010-02-13)
The 4'-(p-toluenesulfonylamino)-4-hydroxychalcone (TSAHC), which bears inhibitory chemotypes for both alpha-glucosidase and tyrosinase, was evaluated for tyrosinase activity and depigmenting ability relative to compounds designed to only target tyrosianse activity. TSAHC emerged to be a competitive reversible inhibitor of mushroom tyrosinase.
Charles Bodet et al.
European journal of medicinal chemistry, 43(8), 1612-1620 (2007-12-21)
The aim of this study was to investigate the effect of 3-(4'-geranyloxy-3'-methoxyphenyl)-2-trans propenoic acid, active principle isolated from Acronychia baueri Schott, and its ester derivatives on biofilm formation by two important oral pathogens, Porphyromonas gingivalis and Streptococcus mutans. The parent
Maya Mori-Hongo et al.
Journal of natural products, 72(1), 63-71 (2008-12-24)
From the roots of Lespedeza cyrtobotrya, 45 flavonoids were isolated along with 20 new and 25 known compounds. Lipophilic flavonoids 2, 3, 7, 9, 11, 28, 30, and 39 exhibited strong inhibitory activities on melanin synthesis in normal human epidermal
Maya Mori-Hongo et al.
Journal of natural products, 72(2), 194-203 (2009-01-10)
In the course of our search for new melanin synthesis inhibitors from plants, 40 new flavonoids and 11 known flavonoids were isolated from the roots of Lespedeza floribunda Bunge. The structures of the new compounds were determined by MS and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service