Skip to Content
Merck
All Photos(1)

Key Documents

239801

Sigma-Aldrich

Picric acid

ReagentPlus®, ≥99%

Synonym(s):

2,4,6-Trinitrophenol, PA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)3C6H2OH
CAS Number:
Molecular Weight:
229.10
Colour Index Number:
10305
Beilstein:
423400
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

1 mmHg ( 195 °C)

product line

ReagentPlus®

Assay

≥99%

contains

35% water

expl. lim.

0.01 %

mp

122-123 °C (dried material) (lit.)

SMILES string

Oc1c(cc(cc1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H3N3O7/c10-6-4(8(13)14)1-3(7(11)12)2-5(6)9(15)16/h1-2,10H

InChI key

OXNIZHLAWKMVMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Desen. Expl. 1

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A G Miriam Leenders et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 28(44), 11333-11346 (2008-10-31)
Ca(v)2.2 channels are localized at nerve terminals where they play a critical role in neurotransmission. However, the determinant that controls surface retention of these channels has not been identified. Here, we report that presynaptic surface localization of Ca(v)2.2 is mediated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service