Skip to Content
Merck
All Photos(1)

Key Documents

238759

Sigma-Aldrich

2-Nitro-4-(trifluoromethyl)phenol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3(CF3)OH
CAS Number:
Molecular Weight:
207.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
99%

Assay

99%

form

liquid

refractive index

n20/D 1.504 (lit.)

bp

92-94 °C/12 mmHg (lit.)

density

1.473 g/mL at 25 °C (lit.)

functional group

fluoro
nitro

SMILES string

Oc1ccc(cc1[N+]([O-])=O)C(F)(F)F

InChI

1S/C7H4F3NO3/c8-7(9,10)4-1-2-6(12)5(3-4)11(13)14/h1-3,12H

InChI key

XZEDEVRSUANQEM-UHFFFAOYSA-N

General description

2-Nitro-4-(trifluoromethyl)phenol is the major product of solution phase photodecomposition of fluorodifen.

Application

2-Nitro-4-(trifluoromethyl)phenol has been used in the synthesis of 2-methyl-2-[2-nitro-4-(trifluoromethyl)phenoxy]propionic acid ethyl ester.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unexpected smiles rearrangement during borane reduction of 2-aryloxy-2-methylpropionamides.
Buchstaller H-P and Anlauf U.
Synthesis, 2005(4), 639-643 (2005)
The aqueous photolysis of TFM and related trifluoromethylphenols. An alternate source of trifluoroacetic acid in the environment.
Ellis DA and Mabury SA.
Environmental Science & Technology, 34(4), 632-637 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service