Skip to Content
Merck
All Photos(3)

Documents

237981

Sigma-Aldrich

Sodium hexafluoroantimonate(V)

technical grade

Synonym(s):

Sodium hexafluoroantimonate(1-), Sodium hexafluorostibanuide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaSbF6
CAS Number:
Molecular Weight:
258.74
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

form

powder and chunks

reaction suitability

reagent type: catalyst
core: antimony

solubility

H2O: slightly soluble(lit.)

density

3.375 g/mL at 25 °C (lit.)

SMILES string

[Na+].F[Sb-](F)(F)(F)(F)F

InChI

1S/6FH.Na.Sb/h6*1H;;/q;;;;;;+1;+5/p-6

InChI key

HKLMYZVMEYYVBS-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuang Chen et al.
Journal of the American Chemical Society, 141(24), 9603-9609 (2019-06-12)
Near infrared (near-IR) electrogenerated chemiluminescence (ECL) from rod-shape bimetallic Au12Ag13 nanoclusters is reported. With ECL standard tris(bipyridine)ruthenium(II) complex (Ru(bpy)3) as reference, the self-annihilation ECL of the Au12Ag13 nanoclusters is about 10 times higher. The coreactant ECL of Au12Ag13 is about
Daniel T Walters et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(15), 3849-3857 (2018-12-24)
Two new molecular boxes, the mono-bromo box [Au6 (Triphos)4 Br](SbF6 )5 ⋅6(CH2 Cl2 ), 4 mB, and the dibromo box, [Au6 (Triphos)4 Br2 ⋅H2 O](SbF6 )4 ⋅4(CH2 Cl2 ), 5 dB, have been prepared in crystalline form. Although constructed from non-luminescent components

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service