Skip to Content
Merck
All Photos(1)

Key Documents

233072

Sigma-Aldrich

4-(Trifluoromethyl)mandelic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4CH(OH)CO2H
CAS Number:
Molecular Weight:
220.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

solid

functional group

carboxylic acid
fluoro
hydroxyl

SMILES string

OC(C(O)=O)c1ccc(cc1)C(F)(F)F

InChI

1S/C9H7F3O3/c10-9(11,12)6-3-1-5(2-4-6)7(13)8(14)15/h1-4,7,13H,(H,14,15)

InChI key

SDGXYUQKJPFLDG-UHFFFAOYSA-N

General description

Bismuth-catalyzed oxidation of 4-(trifluoromethyl)mandelic acid has been reported[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bi0-Catalyzed Oxidation of Mandelic Acid Derivatives: Substrate Selectivity.
Favier I, et al.
European Journal of Organic Chemistry, 2002(12), 1984-1988 (2002)
D B Matthews et al.
Journal of chromatography. B, Biomedical sciences and applications, 695(2), 279-285 (1997-08-01)
Methods for the nuclear magnetic resonance and gas chromatographic analysis of the enantiomers of p-trifluoromethylmandelic acid (p-TFM) and Mosher's acid (alpha-methoxy-alpha-(trifluoromethyl)phenylacetic acid) present in rat urine samples are described. Gas chromartography was performed using cyclodextrin capillary columns with both compounds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service