Skip to Content
Merck
All Photos(1)

Key Documents

232475

Sigma-Aldrich

3,4-Dichlorophenylacetonitrile

98%

Synonym(s):

3,4-Dichlorobenzyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CH2CN
CAS Number:
Molecular Weight:
186.04
Beilstein:
638749
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
98%
To inquire about this product 232475, please contact your local Merck office or dealer. Contact Technical Service

Assay

98%

form

solid

bp

170 °C/12 mmHg (lit.)

mp

39-42 °C (lit.)

functional group

chloro
nitrile

SMILES string

Clc1ccc(CC#N)cc1Cl

InChI

1S/C8H5Cl2N/c9-7-2-1-6(3-4-11)5-8(7)10/h1-2,5H,3H2

InChI key

QWZNCAFWRZZJMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,4-Dichlorophenylacetonitrile was used in the synthesis of:
  • SR 48968, a potent, competitive and selective non-peptide antagonist of the neurokinin A (NK2) receptor[1]
  • tachykinin NK3 receptor antagonist SR 142801, (R)-N-[ 1-[3-[ 1-benzoyl-3-(3,4-dichlororphenyl)-3-piperidinyl]propyyl]-4-phenyl-4-piperidinyl-N-methyl acetamide[2]
  • MDL 105,212, a non-peptide tachykinin receptor antagonist[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacological profile and chemical synthesis of SR 48968, a non-peptide antagonist of the neurokinin A (NK2) receptor.
Emonds-Alt X, et al.
Bioorganic & Medicinal Chemistry Letters, 3(5), 925-930 (1993)
A practical and scalable synthesis of SR 142801, a tachykinin NK3 antagonist.
Chen HG, et al.
Bioorganic & Medicinal Chemistry Letters, 7(5), 555-560 (1997)
Identification and chemical synthesis of MDL 105,212, a non-peptide tachykinin antagonist with high affinity for NK1and NK2 receptors.
Burkholder TP, et al.
Bioorganic & Medicinal Chemistry Letters, 6(8), 951-956 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service