Skip to Content
Merck
All Photos(1)

Key Documents

231258

Sigma-Aldrich

1-Aminohexylamide gel

~2.4 meq/g

Synonym(s):

Poly(acrylic acid 6-aminohexylamide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CH[C(O)NH(CH2)6NH2]-]n
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

description

gel volume in water 4 4mL/g

Quality Level

specific activity

~2.4 meq/g

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: proton reactive

functional group

amine

Application

Affinity chromatography.

Features and Benefits

Cross-linked poly(methyl acrylate) reacted with 1,6-diaminohexane to form amino-amide. Used in water or organic solvents. Stable at pH 2-11. Good chemical and mechanical stability, high terminal amine content, low steric hindrance to amine groups.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service