Skip to Content
Merck
All Photos(1)

Key Documents

229458

Sigma-Aldrich

Antimony(IV) oxide

99.997%

Synonym(s):

Antimony tetroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sb2O4
CAS Number:
Molecular Weight:
307.52
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.997%

density

5.82 g/mL at 25 °C (lit.)

SMILES string

O=[Sb]O[Sb](=O)=O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service