Skip to Content
Merck
All Photos(3)

Key Documents

226963

Sigma-Aldrich

Poly(4-vinylpyridine), cross-linked

2 % cross-linked with divinylbenzene, powder

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€165.00

€165.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€165.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

€165.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

crosslinking

2 % cross-linked with divinylbenzene

SMILES string

n2ccc(cc2)C=C.c1(c(cccc1)C=C)C=C

InChI

1S/C10H10.C7H7N/c1-3-9-7-5-6-8-10(9)4-2;1-2-7-3-5-8-6-4-7/h3-8H,1-2H2;2-6H,1H2

InChI key

HUEZUDXJCGHIHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Useful as an acid scavenger in non-aqueous systems, e.g., in the reaction of acid halides with alcohols or amines, as catalyst in Knoevenagel-type condensations and for the formation of enamines and ketals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service