Skip to Content
Merck
All Photos(1)

Key Documents

225398

Sigma-Aldrich

2-Isopropoxyphenol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOC6H4OH
CAS Number:
Molecular Weight:
152.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.514 (lit.)

bp

100-102 °C/11 mmHg (lit.)

density

1.03 g/mL at 25 °C (lit.)

SMILES string

CC(C)Oc1ccccc1O

InChI

1S/C9H12O2/c1-7(2)11-9-6-4-3-5-8(9)10/h3-7,10H,1-2H3

InChI key

ZNCUUYCDKVNVJH-UHFFFAOYSA-N

General description

2-Isopropoxyphenol is a metabolite of propoxur.

Application

2-Isopropoxyphenol was used as a target chemical in developing a method to measure the urinary phenolic metabolites of pesticides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramagiri Suma et al.
Journal of analytical toxicology, 29(7), 728-733 (2006-01-20)
This research paper describes the development and validation of an analytical method for the simultaneous determination of propoxur and isopropoxy phenol (IPP, a major metabolite) in both blood and urine of rat using reversed-phase high-performance liquid chromatography (HPLC) employing solid-phase
R Brouwer et al.
American journal of industrial medicine, 24(5), 593-603 (1993-11-01)
In eight greenhouses used for carnation culture, workers engaged in harvesting (n = 16), were monitored for dermal and respiratory exposure and urinary excretion of propoxur. Dermal exposure of hands and forearms was estimated from dislodgable foliar residue, using a
J Hardt et al.
Journal of chromatography. B, Biomedical sciences and applications, 723(1-2), 139-145 (1999-03-18)
Human metabolism of the insecticide propoxur yields 2-isopropoxyphenol (IPP) which is excreted conjugated in urine. In this publication a sensitive and selective analytical method is described which permits the determination of IPP as a suitable parameter for biomonitoring. The clean-up
L H Leenheers et al.
Journal of chromatography, 578(2), 189-194 (1992-07-24)
An analytical method for the assessment of the exposure of workers to the pesticide propoxur through biological monitoring has been developed. This study was part of a survey of occupational exposure to pesticides used in greenhouses for the growing of
W J Meuling et al.
The Science of the total environment, 199(1-2), 165-172 (1997-06-20)
A large number of workers in agriculture are exposed daily (through skin contact) to pesticides either directly during mixing and loading or indirectly due to contact. The aim of this study was to investigate the influence of skin moisture on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service