Skip to Content
Merck
All Photos(2)

Key Documents

224235

Sigma-Aldrich

N,N′-Hexamethylene bis(acetamide)

98%

Synonym(s):

N,N′-Diacetyl-1,6-hexanediamine, HMBA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONH(CH2)6NHCOCH3
CAS Number:
Molecular Weight:
200.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

128-129 °C (lit.)

solubility

water: soluble 5%, clear, colorless

functional group

amide

SMILES string

CC(=O)NCCCCCCNC(C)=O

InChI

1S/C10H20N2O2/c1-9(13)11-7-5-3-4-6-8-12-10(2)14/h3-8H2,1-2H3,(H,11,13)(H,12,14)

InChI key

BNQSTAOJRULKNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N′-Hexamethylene bis(acetamide) was used as an inducing agent in obtaining mononuclear cells from the peripheral blood (PB) sample by Ficoll-Hypaque gradient separation[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuguang Zhu et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 23(2), 430-440 (2016-07-22)
New therapeutic approaches are needed for patients with thyroid cancer refractory to radioiodine treatment. An inhibitor of bromodomain and extraterminal domain (BET) proteins, JQ1, shows potent antitumor effects in hematological cancers and solid tumors. To evaluate whether JQ1 is effective
Samuel T Workenhe et al.
Communications biology, 3(1), 645-645 (2020-11-06)
Cancer immunotherapies using monoclonal antibodies to block inhibitory checkpoints are showing durable remissions in many types of cancer patients, although the majority of breast cancer patients acquire little benefit. Human melanoma and lung cancer patient studies suggest that immune checkpoint
Kara G Lassen et al.
PloS one, 7(1), e30176-e30176 (2012-02-01)
Latently infected cells form the major obstacle to HIV eradication. Studies of HIV latency have been generally hindered by the lack of a robust and rapidly deployable cell model that involves primary human CD4 T lymphocytes. Latently infected cell lines
Shailesh K Choudhary et al.
The Journal of infectious diseases, 197(8), 1162-1170 (2008-04-19)
Novel therapeutic approaches are needed to attack persistent proviral human immunodeficiency type 1 (HIV-1) infection. Hexamethylbisacetamide (HMBA), a hybrid bipolar compound, induces expression of the HIV-1 promoter in the long terminal repeat (LTR) region in a Tat-independent manner but mimics
Richard L Thompson et al.
PLoS pathogens, 5(3), e1000352-e1000352 (2009-03-28)
The mechanism controlling the exit from herpes simplex virus latency (HSV) is of central importance to recurrent disease and transmission of infection, yet interactions between host and viral functions that govern this process remain unclear. The cascade of HSV gene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service