Skip to Content
Merck
All Photos(1)

Key Documents

223107

Sigma-Aldrich

Sarcosine hydrochloride

99%

Synonym(s):

N-Methylaminoacetic acid, N-Methylglycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NHCH2CO2H·HCl
CAS Number:
Molecular Weight:
125.55
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

99%

mp

173-175 °C (lit.)

SMILES string

Cl.CNCC(O)=O

InChI

1S/C3H7NO2.ClH/c1-4-2-3(5)6;/h4H,2H2,1H3,(H,5,6);1H

InChI key

WVKIFIROCHIWAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan E McDunn et al.
The Prostate, 73(14), 1547-1560 (2013-07-05)
Current diagnostic techniques have increased the detection of prostate cancer; however, these tools inadequately stratify patients to minimize mortality. Recent studies have identified a biochemical signature of prostate cancer metastasis, including increased sarcosine abundance. This study examined the association of
King Hang Aaron Lau et al.
Langmuir : the ACS journal of surfaces and colloids, 28(46), 16099-16107 (2012-10-30)
Poly(N-substituted glycine) "peptoids" are a class of peptidomimetic molecules receiving significant interest as engineered biomolecules. Sarcosine (i.e., poly(N-methyl glycine)) has the simplest side chain chemical structure of this family. In this Article, we demonstrate that surface-grafted polysarcosine (PSAR) brushes exhibit
Qunyang Xiang et al.
The journal of physical chemistry. A, 116(42), 10276-10284 (2012-09-21)
Aqueous sarcosine salts are fast carbon dioxide (CO(2)) absorbents suitable for use in postcombustion CO(2) capture in coal-fired power plants. We have developed a detailed reaction scheme including all the reactions in the sarcosine-CO(2)-water system. All unknown rate and equilibrium
Giuseppe Lucarelli et al.
The Journal of urology, 190(6), 2271-2277 (2013-05-15)
SPON2 belongs to the F-spondin family of secreted extracellular matrix proteins. It is deregulated in some tumors, including prostate cancer. In this prospective study we assessed the role of serum SPON2 as a biomarker for prostate cancer diagnosis as well
Eri Hara et al.
Biochimica et biophysica acta, 1830(8), 4046-4052 (2013-04-03)
Nanoparticle of Lactosome, which is composed of poly(l-lactic acid)-base depsipeptide with diameter of 35nm, accumulates in solid tumors by the enhanced permeability and retention (EPR) effect. However, a pharmacokinetic alteration of Lactosome was observed when Lactosome was repeatedly administered. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service