Skip to Content
Merck
All Photos(2)

Key Documents

222801

Sigma-Aldrich

4-Nitrobenzyl chloroformate

97%, for peptide synthesis

Synonym(s):

(4-Nitrophenyl)methyl carbonochloridate, (4-Nitrophenyl)methyl chloroformate, 4-Nitrobenzyloxycarbonyl chloride, p-Nitrobenzyloxycarbonyl chloride, p-nitrobenzyl chloroformate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€47.90
25 G
€110.00

€47.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€47.90
25 G
€110.00

About This Item

Linear Formula:
ClCO2CH2C6H4NO2
CAS Number:
Molecular Weight:
215.59
Beilstein:
912446
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

€47.90


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

4-Nitrobenzyl chloroformate, 97%

Assay

97%

reaction suitability

reaction type: Carbonylations

mp

32-34 °C (lit.)

application(s)

peptide synthesis

functional group

chloro
nitro

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(COC(Cl)=O)cc1

InChI

1S/C8H6ClNO4/c9-8(11)14-5-6-1-3-7(4-2-6)10(12)13/h1-4H,5H2

InChI key

MHSGOABISYIYKP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eun Hyang Jang et al.
International journal of pharmaceutics, 580, 119237-119237 (2020-03-24)
Hypoxia is a characteristic feature of various ischemic diseases, including cancer. This study describes the development of glycol chitosan nanoparticles, hydrophobically modified with 4-nitrobenzyl chloroformate and folic acid (FA), that can specifically release drugs under hypoxic conditions. This hypoxia-responsive glycol

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service