Skip to Content
Merck
All Photos(1)

Key Documents

222739

Sigma-Aldrich

2-Phenylbutyronitrile

95%

Synonym(s):

α-Ethylphenylacetonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€47.70

€47.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€47.70

About This Item

Linear Formula:
C6H5CH(CH2CH3)CN
CAS Number:
Molecular Weight:
145.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€47.70


Please contact Customer Service for Availability

Request a Bulk Order

Assay

95%

refractive index

n20/D 1.5086 (lit.)

bp

114-115 °C/15 mmHg (lit.)

density

0.974 g/mL at 25 °C (lit.)

functional group

nitrile
phenyl

SMILES string

CCC(C#N)c1ccccc1

InChI

1S/C10H11N/c1-2-9(8-11)10-6-4-3-5-7-10/h3-7,9H,2H2,1H3

InChI key

IZPUPXNVRNBDSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Phenylbutyronitrile was used in the preparation of Michael-adducts[1]. It was also used in the preparation of S-amides by nitrile hydratase[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

t-BuOK-Catalyzed addition of ketones and nitriles to vinylic silanes, phosphines and thio derivatives.
Bunlaksananusorn T, et al.
Chemical Communications (Cambridge, England), 8, 745-746 (2001)
Enantioselective nitrile hydratases and amidases from different bacterial isolates.
Journal of Molecular Catalysis. B, Enzymatic, 5(1), 137-141 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service