Skip to Content
Merck
All Photos(1)

Key Documents

222097

Sigma-Aldrich

4-Hexylbenzoyl chloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5C6H4COCl
CAS Number:
Molecular Weight:
224.73
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

liquid

refractive index

n20/D 1.5256 (lit.)

density

1.029 g/mL at 25 °C (lit.)

functional group

acyl chloride

SMILES string

CCCCCCc1ccc(cc1)C(Cl)=O

InChI

1S/C13H17ClO/c1-2-3-4-5-6-11-7-9-12(10-8-11)13(14)15/h7-10H,2-6H2,1H3

InChI key

XRAHLPNMIIAEPP-UHFFFAOYSA-N

Application

4-Hexylbenzoyl chloride was used in the synthesis of 6,6′-bis(hexylbenzoylamino)-2,2′-bipyridine (LH2).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nickel (II) Complex of 6, 6'-Bis (hexylbenzoylamino)-2, 2'-bipyridine as a Functional Carrier: pH-Induced Affinity-Switching and Mechanism of the SCN--Selective Up-Hill Transport.
Bulletin of the Chemical Society of Japan, 69(5), 1213-1222 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service