Skip to Content
Merck
All Photos(1)

Key Documents

219746

Sigma-Aldrich

DL-Homocysteic acid

97%

Synonym(s):

DL-2-Amino-4-sulfobutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO3SCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
183.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

273 °C (dec.) (lit.)

solubility

water: soluble 50 mg/mL, clear to slightly hazy, colorless to yellow

functional group

amine
carboxylic acid
sulfonic acid

SMILES string

NC(CCS(O)(=O)=O)C(O)=O

InChI

1S/C4H9NO5S/c5-3(4(6)7)1-2-11(8,9)10/h3H,1-2,5H2,(H,6,7)(H,8,9,10)

InChI key

VBOQYPQEPHKASR-UHFFFAOYSA-N

General description

Complexes of DL-homocysteic acid with Na+, Cu2+, Zn2+ and Ni2+ has been synthesized[1]. DL-Homocysteic acid induces oxidative stress in brain of immature rats[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui-Zhen Jia et al.
Guang pu xue yu guang pu fen xi = Guang pu, 27(7), 1291-1294 (2007-10-20)
The complexes of DL-homocysteic acid (DLH) with Na+, Cu2+, Zn2+ and Ni2+ were synthesized and elemental analyses were used to detect the compositions of these complexes. FTIR spectroscopy was employed to study these coordination structures. The results indicated that all
Tohru Hasegawa et al.
PloS one, 5(1), e8593-e8593 (2010-01-26)
Alzheimer's disease (AD) is an age-associated progressive neurodegenerative disorder with dementia, the exact pathogenic mechanisms of which remain unknown. We previously reported that homocysteic acid (HA) may be one of the pathological biomarkers in the brain with AD and that
Marshall Bern et al.
Rapid communications in mass spectrometry : RCM, 24(6), 768-772 (2010-02-20)
Analysis of protein oxidation is necessary in numerous areas of biochemistry, including hydroxyl radical surface mapping, oxidative stress assays, and pharmaceutical stability testing. Mass spectrometry is one of the tools most often used to identify protein oxidation products, and previous
Hari H Subramanian et al.
The Journal of comparative neurology, 521(13), 3083-3098 (2013-05-01)
The midbrain periaqueductal gray (PAG) is involved in many basic survival behaviors that affect respiration. We hypothesized that the PAG promotes these behaviors by changing the firing of preinspiratory (pre-I) neurons in the pre-Bötzinger complex, a cell group thought to
Peng Li et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 299(5), R1369-R1376 (2010-08-20)
We have shown that electroacupuncture (EA) inhibits sympathoexcitatory rostral ventrolateral medulla (rVLM) neurons and reflex responses following activation of a long-loop pathway in the arcuate nucleus and ventrolateral periaqueductal gray (vlPAG). Additionally, EA at P 5-6 acupoints (overlying the median

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service