Skip to Content
Merck
All Photos(1)

Key Documents

219592

Sigma-Aldrich

2-(1,3-Dioxan-2-yl)ethyltriphenylphosphonium bromide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26BrO2P
CAS Number:
Molecular Weight:
457.34
Beilstein:
4117571
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

98%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

193-195 °C (lit.)

functional group

ether
phosphine

SMILES string

[Br-].C1COC(CC[P+](c2ccccc2)(c3ccccc3)c4ccccc4)OC1

InChI

1S/C24H26O2P.BrH/c1-4-11-21(12-5-1)27(22-13-6-2-7-14-22,23-15-8-3-9-16-23)20-17-24-25-18-10-19-26-24;/h1-9,11-16,24H,10,17-20H2;1H/q+1;/p-1

InChI key

XETDBHNHTOJWPZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

2-(1,3-Dioxan-2-yl)ethyltriphenylphosphonium bromide can be used:
  • As a three-carbon homologating agent used to prepare α,β- or β,γ- unsaturated compounds.[1]
  • In the olefination of methyl 5-oxopentanoate to yield methyl 7-(1,3-dioxan2-yl)hept-(5Z)-enoate.[2]
  • As a cathode interfacial layer material in polymer solar cells.[3]

Reactant for:
  • Wittig olefination

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organophosphorus Derivatives as Cathode Interfacial-Layer Materials for Highly Efficient Fullerene-Free Polymer Solar Cells
Gupta M, et al.
ACS Applied Materials & Interfaces, 10(42), 35896-35903 (2018)
2-(1, 3-Dioxan-2-yl)-ethylidenetriphenylphosphorane, A New Three-Carbon Homologating Agent
Stowell JC and Keith DR
Synthesis, 1979(2), 32-133 (1979)
Algal pheromone biosynthesis: stereochemical analysis and mechanistic implications in gametes of Ectocarpus siliculosus
Rui F and Boland W
The Journal of Organic Chemistry, 75(12), 3958-3964 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service