Skip to Content
Merck
All Photos(1)

Key Documents

218855

Sigma-Aldrich

9-Chloro-9-phenylxanthene

97%

Synonym(s):

Pixyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H13ClO
CAS Number:
Molecular Weight:
292.76
Beilstein:
237712
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

102-106 °C (lit.)

solubility

chloroform: soluble 25 mg/mL, clear, yellow to orange

SMILES string

ClC2(c1ccccc1)c3ccccc3Oc4ccccc24

InChI

1S/C19H13ClO/c20-19(14-8-2-1-3-9-14)15-10-4-6-12-17(15)21-18-13-7-5-11-16(18)19/h1-13H

InChI key

HTGMODSTGYKJDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

9-Chloro-9-phenylxanthene was used in the preparation of (1S,5S,6R,8S)-6-[(allyloxycarbonyl)oxy]-5-(trifluoroacetamido)-8-[9′-(9-phenylxanthenyl)oxy]-2-oxabicyclo(3.2.1)octane[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The pixyl (Px) group: a novel photocleavable protecting group for primary alcohols.
Misetic A and Boyd MK.
Tetrahedron Letters, 39(13), 1653-1656 (1998)
I Pompizi et al.
Nucleic acids research, 28(14), 2702-2708 (2000-07-25)
The synthesis and incorporation into oligodeoxy-nucleotides of two novel, conformationally restricted abasic (AB) site analogs are described. The stability of oligonucleotide 18mer duplexes containing one such AB site opposite any of the four natural DNA bases was investigated by UV
The 9-phenylxanthen-9-yl protecting group.
Chattopadhyaya JB and Reese CB.
Journal of the Chemical Society. Chemical Communications, 15, 639-640 (1978)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service