Skip to Content
Merck
All Photos(1)

Key Documents

214337

Sigma-Aldrich

4-Fluoro-3-nitrobenzotrifluoride

99%

Synonym(s):

3-Nitro-α,α,α,4-tetrafluorotoluene, 4,α,α,α-Tetrafluoro-3-nitrotoluene, FNBT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(NO2)CF3
CAS Number:
Molecular Weight:
209.10
Beilstein:
1880508
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
99%

Assay

99%

refractive index

n20/D 1.462 (lit.)

bp

92 °C/15 mmHg (lit.)

density

1.494 g/mL at 25 °C (lit.)

functional group

fluoro
nitro

SMILES string

[O-][N+](=O)c1cc(ccc1F)C(F)(F)F

InChI

1S/C7H3F4NO2/c8-5-2-1-4(7(9,10)11)3-6(5)12(13)14/h1-3H

InChI key

HLDFCCHSOZWKAA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Fluoro-3-nitrobenzotrifluoride was used in pre-column derivatization technique for HPLC with UV/VIS spectrophotometric detection of polyamines[1].
It was also used in the synthesis of:
  • 2-nitro-4-trifluoromethylphenylhydrazine, derivatization reagent for 1,2-diol compounds and oxo-steroids[2]
  • [3-(2-nitro-4-trifluoromethylphenyl)aminophenyl]dihydroxyborane, derivatization reagent for 1,2-diol compounds and oxo-steroids[2]
  • 1-(3-chlorophenyl)-5-trifluoromethyl-3-hydrobenzimidazol-2-one[3]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Molins-Legua et al.
The Analyst, 124(4), 477-482 (1999-12-22)
The derivatization of biogenic amines such as putrescine, cadaverine, spermidine and spermine with dansyl chloride in solid phase extraction cartridges is described. Different types of filling materials were tested in order to have the highest retention of the different analytes.
Tatsuya Higashi et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 18(12), 1301-1307 (2002-12-28)
Derivatization of neutral steroids for increasing sensitivity in liquid chromatography/negative atmospheric pressure chemical ionization-mass spectrometry (APCI-MS) has been examined. Under APCI conditions, gas-phase electrons are provided by the corona discharge and captured by electron-affinitive compounds. In negative APCI-MS, therefore, ultrahigh
Emanuela Caci et al.
American journal of physiology. Lung cellular and molecular physiology, 285(1), L180-L188 (2003-03-26)
Activators of the CFTR Cl- channel may be useful for therapy of cystic fibrosis. Short-circuit current (Isc) measurements were done on human bronchial epithelial cells to characterize the best flavone and benzimidazolone CFTR activators identified by lead-based combinatorial synthesis and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service