Skip to Content
Merck
All Photos(1)

Key Documents

213195

Sigma-Aldrich

Lithium acetate dihydrate

98%

Synonym(s):

Acetic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOLi · 2H2O
CAS Number:
Molecular Weight:
102.02
Beilstein:
3564320
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
solid
Pricing and availability is not currently available.

grade

for analytical purposes

Assay

98%

form

solid

mp

53-56 °C (lit.)

SMILES string

[Li+].[H]O[H].[H]O[H].CC([O-])=O

InChI

1S/C2H4O2.Li.2H2O/c1-2(3)4;;;/h1H3,(H,3,4);;2*1H2/q;+1;;/p-1

InChI key

IAQLJCYTGRMXMA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Crystals of lithium acetate dihydrate belong to the orthorhombic crystal system.[1] On irradiation CH3COOLi.2H2O crystals generates OH radicals. ESR study reports the g-tensor and hyperfine tensor values as (2.0038, 2.0071, 2.0457) and (-22.3, -45.8, +7.2G), respectively.[2]

Application

Lithium acetate dihydrate (CH3COOLi.2H2O) may be used in the preparation of the solution of lithium acetate[3] and single crystals of lithium acetate dihydrate.[4]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ESR study of OH radicals in irradiated lithium acetate dihydrate single crystals.
Premovic P and Gal O.
Journal of Magnetic Resonance, 13(2), 177-180 (1974)
The crystal structure of lithium acetate dihydrate CH3COOLi.2H2O.
Amirthalingam VT and Padmanabhan VM.
Acta Crystallographica, 11(12), 896-896 (1958)
Lithium nuclear-magnetic-resonance in lithium acetate dihydrate, CH3COOLi. 2H2O.
Bhat SV, et al.
Acta Crystallographica Section B, Structural Science, 30(4), 846-848 (1974)
Transformation of Saccharomyces cerevisiae by the lithium acetate/single-stranded carrier DNA/polyethylene glycol protocol.
Agatep R, et al.
Technical Tips Online, 3(1), 133-137 (1998)
Zico Alaia Akbar et al.
Optics express, 23(19), A1280-A1287 (2015-09-26)
A unique, hierarchically structured, aggregated TiO(2) nanowire (A-TiO(2)-nw) is prepared by solvothermal synthesis and used as a dual-functioning photoelectrode in dye-sensitized solar cells (DSSCs). The A-TiO(2)-nw shows improved light scattering compared to conventional TiO(2) nanoparticles (TiO(2)-np) and dramatically enhanced dye

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service