Skip to Content
Merck
All Photos(1)

Documents

208795

Sigma-Aldrich

S-Methyl methanethiosulfonate

97%

Synonym(s):

S-Methyl thiomethanesulfonate, MMTS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO2SCH3
CAS Number:
Molecular Weight:
126.20
Beilstein:
1446059
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.513 (lit.)

bp

69-71 °C/0.4 mmHg (lit.)

solubility

DMF: soluble 1:1
chloroform: soluble 200 ul per mL, clear, faintly yellow-green
water: soluble 1:5

density

1.337 g/mL at 25 °C (lit.)

SMILES string

CSS(C)(=O)=O

InChI

1S/C2H6O2S2/c1-5-6(2,3)4/h1-2H3

InChI key

XYONNSVDNIRXKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

S-Methyl methanethiosulfonate is a reagent used to synthesize thiols and thioesters. Used as cross-linking agent to prepare polymer networks. Interaction of S-methyl methanethiosulfonate (MMTS) with dipalmitoylphosphatidylcholine (DPPC) bilayers has been investigated by FTIR and surface-enhanced Raman spectroscopy.

Application

S-Methyl methanethiosulfonate was used as sulfenylating agent for β-keto sulfoxides, methylene compounds, half-esters of malonic acids and aryl Grignard reagents. It was also used as a reagent to trap the natural thiol-disulfide state of the proteins.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Perkin Transactions 1, 3167-3167 (1993)
Sustainable Three-Component Synthesis of Isothioureas from Isocyanides, Thiosulfonates, and Amines
P Mampuys, et al.
Angewandte Chemie (International Edition in English), 126 (2014)
Sustainable Three-Component Synthesis of Isothioureas from Isocyanides, Thiosulfonates, and Amines
P Mampuys, et al.
Angewandte Chemie (International Edition in English), 126, 13063-13068 (2014)
Synthetic Communications, 22, 1359-1359 (1992)
The Journal of Organic Chemistry, 58, 6132-6132 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service