Skip to Content
Merck
All Photos(1)

Key Documents

206350

Sigma-Aldrich

1-Pyrrolidinecarbonyl chloride

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€50.50
5 G
€76.10

€50.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€50.50
5 G
€76.10

About This Item

Empirical Formula (Hill Notation):
C5H8ClNO
CAS Number:
Molecular Weight:
133.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€50.50


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.493 (lit.)

bp

104-106 °C/14 mmHg (lit.)

density

1.209 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

−20°C

SMILES string

ClC(=O)N1CCCC1

InChI

1S/C5H8ClNO/c6-5(8)7-3-1-2-4-7/h1-4H2

InChI key

XACWJIQLDLUFSR-UHFFFAOYSA-N

Application

1-Pyrrolidinecarbonyl chloride was used in the synthesis of (S)-4-(2-(((benzyloxy)carbonyl)amino)-3-methoxy-3-oxopropyl)phenyl pyrrolidine-1-carboxylate[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin Cao et al.
Organic & biomolecular chemistry, 12(6), 965-978 (2013-12-24)
The α9β1 and α4β1 integrin subtypes are expressed on bone marrow haemopoietic stem cells and have important roles in stem cell regulation and trafficking. Although the roles of α4β1 integrin have been thoroughly investigated with respect to HSC function, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service