Skip to Content
Merck
All Photos(3)

Key Documents

205923

Sigma-Aldrich

Platinum on carbon

greener alternative

extent of labeling: 1 wt. % loading, matrix activated carbon support

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

form

powder

reaction suitability

reagent type: catalyst
core: platinum

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

1 wt. % loading

matrix

activated carbon support

greener alternative category

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Platinum on carbon (Pt/C) can be used as a catalyst for a variety of applications, such as:
  • hydrosilylation reaction for the production of molecules with silicon-carbon bonds[1]
  • direct alcohol fuel cells (DAFCs)[2][3]
  • an electrocatalyst in hydrogen evolution reaction (HER)[4]
  • formation of nanocatalyst for polymer electrolyte fuel cells[5]

Other Notes

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly efficient hydrogen evolution from seawater by a low-cost and stable CoMoP@ C electrocatalyst superior to Pt/C
Ma Y, et al.
Energy & Environmental Science, 10(3), 788-798 (2017)
Synthesis and physical/electrochemical characterization of Pt/C nanocatalyst for polymer electrolyte fuel cells
Prabhuram J, et al.
Journal of Power Sources, 134(1), 1-6 (2004)
Ethanol oxidation reactions using SnO2@ Pt/C as an electrocatalyst
Silva JCM, et al.
Applied Catalysis. B, Environmental, 99(1-2), 265-271 (2010)
Ethanol electrooxidation on Pt/C and Pd/C catalysts promoted with oxide
Xu C, et al.
Journal of Power Sources, 164(2), 527-531 (2007)
Hydrosilylation of alkynes catalyzed by platinum on carbon
Chauhan M, et al.
Journal of Organometallic Chemistry, 645(1-2), 1-13 (2002)

Articles

Polymer electrolyte membrane (PEM) fuel cells offer high efficiency and low-temperature operation, reducing oil use in light-duty vehicle transportation.

Proton exchange membrane (PEM) fuel cells operate at relatively low temperatures and are composed of two electrodes and a conductive elecrolyte.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service