203971
Palladium(II) oxide
99.995% trace metals basis
Synonym(s):
Palladium monoxide, Palladium oxide, Palladium(2+) oxide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
OPd
CAS Number:
Molecular Weight:
122.42
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay
99.995% trace metals basis
form
powder
reaction suitability
reagent type: catalyst
core: palladium
mp
870 °C (lit.)
density
8.7 g/mL at 25 °C (lit.)
SMILES string
O=[Pd]
InChI
1S/O.Pd
InChI key
HBEQXAKJSGXAIQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
AB2O4 phase, LuPd2O4, with potential uses in superconducting material as well as being of geochemical importance, was formed at elevated pressure and temperature from Lu2O3 and PdO in the presence of potassium chlorate.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Ox. Sol. 2
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Self-assembly of a heteropolyoxopalladate nanocube: [Pd(II)13As(V)8O34(OH)6]8-.
Elena V Chubarova et al.
Angewandte Chemie (International ed. in English), 47(49), 9542-9546 (2008-10-14)
Masanao Yokohira et al.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie, 58(4), 211-221 (2006-11-25)
We have established and documented an in vivo bioassay for detection of hazards with intratracheally instilled fine particles, which can be used for risk assessment of toxicity of materials inhaled into deep lung tissue of humans (Yokohira et al. Establishment
Qian Liu et al.
Journal of nanoscience and nanotechnology, 7(1), 362-367 (2007-04-26)
The optical reflectance and transmittance of Platinum oxide (PtO(x)) and palladium oxide (PdO(x)) thin films for ultrahigh-density optical storage are investigated using Z-scan technique under irradiation of blue laser (442 nm). The power thresholds of the PtO(x) and PdO(x) decomposition
Qi Li et al.
Environmental science & technology, 43(5), 1534-1539 (2009-04-09)
Removal of arsenic species from water by palladium-modified nitrogen-doped titanium oxide (TiON/PdO) nanoparticles was investigated with and without visible light. For the first time, a high degree of As(III) removal undervisible light illumination was demonstrated on oxide photocatalysts. Over 2
Qi Li et al.
Environmental science & technology, 44(9), 3493-3499 (2010-04-15)
A novel composite photocatalyst of nitrogen (N) and fluorine (F) codoped titanium oxide (TiO(2)) nanotube arrays with dispersed palladium oxide (PdO) nanoparticles was developed by dispersing PdO nanoparticles into N and F codoped TiO(2) nanotube array template created by anodization
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service