201650
Fluorescamine
98%
Synonym(s):
4-Phenylspiro-[furan-2(3H),1-phthalan]-3,3′-dione
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C17H10O4
CAS Number:
Molecular Weight:
278.26
Beilstein:
921143
EC Number:
MDL number:
UNSPSC Code:
12352204
Recommended Products
Assay
98%
mp
153-157 °C (lit.)
λmax
306 nm
SMILES string
O=C1OC2(OC=C(C2=O)c3ccccc3)c4ccccc14
InChI
1S/C17H10O4/c18-15-13(11-6-2-1-3-7-11)10-20-17(15)14-9-5-4-8-12(14)16(19)21-17/h1-10H
InChI key
ZFKJVJIDPQDDFY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Non-fluorescent reagent that reacts readily under mild conditions with primary amines in amino acids and peptides to form stable, highly fluorescent compounds. Low background due to hydrolysis. Useful for the fluorometric assay of amino acids, protein, and proteolytic enzymes. Effectively blocks newly generated amino termini in protein sequence analyses.
replaced by
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Claire E Stanley et al.
Chimia, 66(3), 88-98 (2012-05-02)
This account highlights some of our recent activities focused on developing microfluidic technologies for application in high-throughput and high-information content chemical and biological analysis. Specifically, we discuss the use of continuous and segmented flow microfluidics for artificial membrane formation, the
Khin Yin Win et al.
The Analyst, 137(10), 2328-2332 (2012-03-13)
Here we report a fast and effective method to visualize interactive proteins across intact mammalian cells via on-site formation of fluorescence using instant reaction of non-fluorescent fluorescamine with primary amines on proteins. Without interference by fluorescence background, this fluorogenic labelling
Andrew D Aubrey et al.
Astrobiology, 8(3), 583-595 (2008-08-06)
The Urey organic and oxidant detector consists of a suite of instruments designed to search for several classes of organic molecules in the martian regolith and ascertain whether these compounds were produced by biotic or abiotic processes using chirality measurements.
Yusuke Suzuki et al.
Journal of agricultural and food chemistry, 56(22), 10811-10816 (2008-11-06)
Photoinduced decarboxylation via homolytic cleavage of the ester linkage generating two benzyl radicals being recoupled is known to be a major photolytic pathway of the insecticide fenvalerate in aqueous or organic solvents. A highly sensitive and selective fluorescence spectroscopic method
Laura Contreras-Ruiz et al.
Cornea, 29(5), 550-558 (2010-03-26)
Hyaluronic acid-chitosan nanoparticles (HA-CS NPs) have the potential to serve as a reliable drug delivery system to topically treat ocular surface disorders. We evaluated the in vivo uptake by ocular structures, the acute tolerance, and possible alterations of tear film
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service