Skip to Content
Merck
All Photos(1)

Key Documents

201324

Sigma-Aldrich

Rhodamine 6G

Dye content 95 %

Synonym(s):

Basic Red 1

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C28H31N2O3Cl
CAS Number:
Molecular Weight:
479.01
Colour Index Number:
45160
Beilstein:
3900071
EC Number:
MDL number:
UNSPSC Code:
12171500

composition

Dye content, 95%

λmax

524 nm

SMILES string

Cl.CCNc1cc2OC3=CC(=N/CC)\C(C)=CC3=C(c2cc1C)c4ccccc4C(=O)OCC

InChI

1S/C28H30N2O3.ClH/c1-6-29-23-15-25-21(13-17(23)4)27(19-11-9-10-12-20(19)28(31)32-8-3)22-14-18(5)24(30-7-2)16-26(22)33-25;/h9-16,29H,6-8H2,1-5H3;1H/b30-24+;

InChI key

VYXSBFYARXAAKO-WTKGSRSZSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A rhodamine analog useful in Pgp efflux assays. Has been used to characterize kinetics of MRP1- mediated efflux.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruno de Gennaro et al.
Colloids and surfaces. B, Biointerfaces, 130, 101-109 (2015-04-29)
In view of zeolite potentiality as a carrier for sustained drug release, a clinoptilolite-rich rock from California (CLI_CA) was superficially modified with cetylpyridinium chloride and loaded with diclofenac sodium (DS). The obtained surface modified natural zeolites (SMNZ) were characterized by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service