Skip to Content
Merck
All Photos(3)

Key Documents

200409

Sigma-Aldrich

Polycaprolactone triol

average Mn ~900

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C[CH2O[CO(CH2)5O]nH]3
CAS Number:
MDL number:
UNSPSC Code:
12162002

mol wt

average Mn ~900

refractive index

n20/D 1.476

transition temp

softening point 30 °C

density

1.08 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

424.4 °F - closed cup

Flash Point(C)

218 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

In the past two decades, tissue engineering and regenerative medicine have become important interdisciplinary fields that span biology, chemistry, engineering, and medicine.

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service