Skip to Content
Merck
All Photos(1)

Documents

197874

Sigma-Aldrich

3-(3,4,5-Trimethoxyphenyl)propionic acid

≥98%

Synonym(s):

3,4,5-Trimethoxyhydrocinnamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2CH2CH2CO2H
CAS Number:
Molecular Weight:
240.25
Beilstein:
1471912
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

powder

mp

100-104 °C (lit.)

SMILES string

COc1cc(CCC(O)=O)cc(OC)c1OC

InChI

1S/C12H16O5/c1-15-9-6-8(4-5-11(13)14)7-10(16-2)12(9)17-3/h6-7H,4-5H2,1-3H3,(H,13,14)

InChI key

ZCYXGVJUZBKJAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service