Skip to Content
Merck
All Photos(1)

Documents

196827

Sigma-Aldrich

2,3,5,6-Tetrafluorotoluene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC6F4CH3
CAS Number:
Molecular Weight:
164.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.42 (lit.)

bp

125-126 °C (lit.)

density

1.347 g/mL at 25 °C (lit.)

SMILES string

Cc1c(F)c(F)cc(F)c1F

InChI

1S/C7H4F4/c1-3-6(10)4(8)2-5(9)7(3)11/h2H,1H3

InChI key

POMGTQLCZJZYAM-UHFFFAOYSA-N

General description

Direct arylation reaction of 2,3,5,6-tetrafluorotoluene with various arylhalides has been investigated.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

73.4 °F - closed cup

Flash Point(C)

23 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc Lafrance et al.
Journal of the American Chemical Society, 128(27), 8754-8756 (2006-07-06)
Penta-, tetra-, tri-, and difluorobenzenes undergo direct arylation with a wide range of arylhalides in high yield. Inverse reactivity is observed compared to the common electrophilic aromatic substitution pathway since electron-deficient, C-H acidic arenes react preferentially. Computational studies indicate that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service