Skip to Content
Merck
All Photos(2)

Key Documents

196258

Sigma-Aldrich

4-Chloro-2-nitrobenzyl chloride

98%

Synonym(s):

α,4-Dichloro-2-nitrotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)CH2Cl
CAS Number:
Molecular Weight:
206.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

38-44 °C (lit.)

functional group

chloro
nitro

SMILES string

[O-][N+](=O)c1cc(Cl)ccc1CCl

InChI

1S/C7H5Cl2NO2/c8-4-5-1-2-6(9)3-7(5)10(11)12/h1-3H,4H2

InChI key

OMPHLGROCARZOU-UHFFFAOYSA-N

Application

4-Chloro-2-nitrobenzyl chloride has been used in the synthesis of 3-iodothyronamine[1].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A New Efficient Synthetic Method for 3-Iodothyronamine and Its Potent Hypothermic Efficacy.
Kim J-G, et al.
Chin. J. Chem., 29(10), 2205-2208 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service