Skip to Content
Merck
All Photos(1)

Key Documents

195871

Sigma-Aldrich

6-Methyl-5-hepten-2-ol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2CH(OH)CH3
CAS Number:
Molecular Weight:
128.21
Beilstein:
1720072
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Assay

99%

form

liquid

refractive index

n20/D 1.448 (lit.)

bp

78 °C/14 mmHg (lit.)

density

0.844 g/mL at 25 °C (lit.)

SMILES string

CC(O)CC\C=C(\C)C

InChI

1S/C8H16O/c1-7(2)5-4-6-8(3)9/h5,8-9H,4,6H2,1-3H3

InChI key

OHEFFKYYKJVVOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenji Mori
Chirality, 23(6), 449-462 (2011-06-03)
Mori's synthetic works on bioactive natural products in general and pheromones in particular started about 40 years ago to establish their absolute configurations and also to clarify their stereochemistry-bioactivity relationships. Results indicate that bioactive natural products are not always enantiomerically
A E Janssen et al.
Enzyme and microbial technology, 18(5), 340-346 (1996-04-01)
The fatty acid specificity of lipase from Candida rugosa during the esterification of saturated fatty acids and sulcatol in toluene has been studied. The true kinetic parameters are obtained by fitting the experimental data to a Ping-Pong kinetic model that
Shui-Ling Chen et al.
Organic letters, 6(19), 3365-3367 (2004-09-10)
[reaction: see text] A highly efficient chemical kinetic resolution of bishomoallylic alcohols was developed when the alcohols underwent In(OTf)(3)-catalyzed 3,5-oxonium-ene-type cyclization with steroidal aldehyde 2. Consistently high enantiomeric excess (up to >99%) was obtained.
Michela Vittorini et al.
Bioprocess and biosystems engineering, 34(2), 247-251 (2010-11-04)
The activity of Thermoanaerobium brockii alcohol dehydrogenase (TBADH) adsorbed on mesoporous silica SBA-15 was compared with that of the free enzyme in water and in biphasic system (water phase up to 50% v/v water). TBADH was active at a water
Paola Gonzalez Audino et al.
Journal of economic entomology, 98(5), 1506-1509 (2005-12-13)
We report here the identification and behavioral activity of volatile compounds emitted by male Platypus mutatus (=sulcatus) Chapuis while boring galleries in living poplar, Populus deltoides Marshall, trees. Headspace analysis using solid phase microextraction techniques showed the presence of 6-methyl-5-hepten-2-ol

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service