Skip to Content
Merck
All Photos(1)

Key Documents

195170

Sigma-Aldrich

Mordant Brown 1

Dye content 40 %

Synonym(s):

5-[2,4-Diamino-5-(2-hydroxy-5-nitrophenylazo)phenylazo]-1-naphthalenesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2N)2C6H2[N=NC6H3(OH)NO2]N=NC10H6SO3Na
CAS Number:
Molecular Weight:
529.46
Colour Index Number:
20110
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 40%

λmax

373 nm
487 nm (2nd)

SMILES string

[Na+].Nc1cc(N)c(cc1N=Nc2cc(ccc2O)[N+]([O-])=O)N=Nc3cccc4c(cccc34)S([O-])(=O)=O

InChI

1S/C22H17N7O6S.Na/c23-15-10-16(24)19(27-28-20-9-12(29(31)32)7-8-21(20)30)11-18(15)26-25-17-5-1-4-14-13(17)3-2-6-22(14)36(33,34)35;/h1-11,30H,23-24H2,(H,33,34,35);/q;+1/p-1

InChI key

KHXQQMBFMDTPSD-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service