Skip to Content
Merck
All Photos(1)

Key Documents

192996

Sigma-Aldrich

α-Methyl-2-naphthalenemethanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH(CH3)OH
CAS Number:
Molecular Weight:
172.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

73-78 °C (lit.)

functional group

hydroxyl

SMILES string

CC(O)c1ccc2ccccc2c1

InChI

1S/C12H12O/c1-9(13)11-7-6-10-4-2-3-5-12(10)8-11/h2-9,13H,1H3

InChI key

AXRKCRWZRKETCK-UHFFFAOYSA-N

General description

α-Methyl-2-naphthalenemethanol forms complexes and multimers with methyl lactate[1]. α-Methyl-2-naphthalenemethanol on reaction with sulfur trioxide-dimethylformamide complex and pyridine yields sulfated products[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiye Tang et al.
Journal of chemical theory and computation, 13(5), 2230-2244 (2017-04-19)
We introduce a novel method, Pathway Search guided by Internal Motions (PSIM), that efficiently finds molecular dissociation pathways of a ligand-receptor system with guidance from principal component (PC) modes obtained from molecular dynamics (MD) simulations. Modeling ligand-receptor dissociation pathways can
Identification of sulfation sites of metabolites and prediction of the compounds' biological effects.
Yi L, et al.
Analytical and Bioanalytical Chemistry, 383(6), 666-674 (2006)
Akiyuki Ryoki et al.
Journal of chromatography. A, 1599, 144-151 (2019-04-21)
Coated-type chiral stationary phases (CSPs) for high-performance liquid chromatography (HPLC) were prepared from three cyclic amylose tris(3,5-dimethylphenylcarbamate) (cADMPC) samples, of which weight-average molar mass (Mw) ranges from 19 to 91 kg mol-1, and from three linear ADMPC samples ranging in Mw
K Le Barbu-Debus et al.
Physical chemistry chemical physics : PCCP, 11(35), 7589-7598 (2009-12-03)
Complexation between (1R,2S)-(+)-cis-1-amino-2-indanol (AI) and the two enantiomers of methyl lactate has been studied by means of laser-induced fluorescence, resonance-enhanced two-photon ionisation, and IR-UV double resonance spectroscopy, in the region of 3 microm. Two isomeric complexes have been spectroscopically characterised
Danlei Wang et al.
Chemico-biological interactions, 315, 108905-108905 (2019-11-26)
Mineral oils are widely applied in food production and processing and may contain polycyclic aromatic hydrocarbons (PAHs). The PAHs that may be present in mineral oils are typically alkylated, and have been barely studied. Metabolic oxidation of the aromatic ring

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service