Skip to Content
Merck
All Photos(1)

Key Documents

192643

Sigma-Aldrich

1-Ethyl-3-hydroxypyridinium bromide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10BrNO
CAS Number:
Molecular Weight:
204.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

chunks

mp

104-108 °C (lit.)

SMILES string

[Br-].CC[n+]1cccc(O)c1

InChI

1S/C7H9NO.BrH/c1-2-8-5-3-4-7(9)6-8;/h3-6H,2H2,1H3;1H

InChI key

ZGQVRFVHWWFKRP-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and gas chromatographic evaluation of a high-temperature hydrogen-bond acid stationary phase.
Martin SD, et al.
Journal of Chromatography A, 805(1), 217235-217235 (1998)
Chemometric evaluation of the solvent properties of liquid organic salts.
Pool SK and Poole CF.
Analyst, 120(2), 289-294 (1995)
Avi Weissberg et al.
Journal of chromatography. A, 1512, 71-77 (2017-07-18)
A methodology for sensitive determination of sarin (GB), soman (GD) and cyclosarin (GF) chemical warfare agents in aqueous media was developed. The method incorporates direct derivatization with 2-[(dimethylamino)methyl]phenol (2-DMAMP), a commercially available, water-soluble reagent, followed by LC-ESI-MS/MS analysis in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service