Skip to Content
Merck
All Photos(1)

Key Documents

190691

Sigma-Aldrich

3-Fluorobenzamide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CONH2
CAS Number:
Molecular Weight:
139.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

solid

mp

129-132 °C (lit.)

SMILES string

NC(=O)c1cccc(F)c1

InChI

1S/C7H6FNO/c8-6-3-1-2-5(4-6)7(9)10/h1-4H,(H2,9,10)

InChI key

YPIGHNIIXYSPKF-UHFFFAOYSA-N

Application

3-Fluorobenzamide has been used in the synthesis of N-(3-fluorobenzoyl)-N′,N′′-bis(4-methylphenyl)phosphoric triamide[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehrdad Pourayoubi et al.
Acta crystallographica. Section E, Structure reports online, 67(Pt 11), o3034-o3034 (2012-01-06)
In the title compound, C(21)H(21)FN(3)O(2)P, the NH and P(=O) groups of the C(=O)NHP(=O) fragment are in a syn arrangement with respect to each other, as are the two amide H atoms of the two CH(3)-4-C(6)H(4)-NH moieties. In the crystal, mol-ecules

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service