Skip to Content
Merck
All Photos(1)

Key Documents

189308

Sigma-Aldrich

4-Bromophenethylamine

98%

Synonym(s):

2-(4-Bromophenyl)ethan-1-amine, 2-(4-Bromophenyl)ethanamine, 2-(4-Bromophenyl)ethylamine, 4-Bromobenzeneethanamine, 4-Bromophenylethylamine, p-Bromophenethylamine, p-Bromophenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH2CH2NH2
CAS Number:
Molecular Weight:
200.08
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.574 (lit.)

bp

63-72 °C/0.2 mmHg (lit.)

density

1.29 g/mL at 25 °C (lit.)

functional group

amine
bromo

SMILES string

NCCc1ccc(Br)cc1

InChI

1S/C8H10BrN/c9-8-3-1-7(2-4-8)5-6-10/h1-4H,5-6,10H2

InChI key

ZSZCXAOQVBEPME-UHFFFAOYSA-N

Application

4-Bromophenethylamine was used in the synthesis of pyrazinoisoquinoline derivatives and N-2-(4-bromophenyl)ethyl chloroacetamide[1]. It was also used in the synthesis of alkyl arylamino sufides employing elemental sulfur and various halides[2].
Used in a synthesis of alkyl arylamino sufides employing elemental sulfur and a variety of halides (benzyl, allyl, primary, etc).[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark Eggink et al.
Analytical and bioanalytical chemistry, 397(2), 665-675 (2010-03-20)
Based on the template of a recently introduced derivatization reagent for aldehydes, 4-(2-(trimethylammonio)ethoxy)benzeneaminium dibromide (4-APC), a new derivatization agent was designed with additional features for the analysis and screening of biomarkers of lipid peroxidation. The new derivatization reagent, 4-(2-((4-bromophenethyl)dimethylammonio)ethoxy)benzenaminium dibromide
Jungyeob Ham et al.
The Journal of organic chemistry, 71(15), 5781-5784 (2006-07-15)
We have developed two simple and high yielding one-pot syntheses of alkyl aminoaryl sulfides containing a series of four-steps: in situ protection of the free amine by reaction with a Grignard reagent, halogen-lithium exchange, sulfur insertion, and a substitution reaction
Formation of pyrazinoisoquinoline ring system by the tandem amidoalkylation and N-acyliminium ion cyclization: An efficient synthesis of Praziquantel.
Kim JH, et al.
Tetrahedron, 54(26), 7395-7400 (1998)
Boyoung Han et al.
Drug testing and analysis, 11(3), 382-391 (2018-09-07)
Target analysis using liquid chromatography-tandem mass spectrometry is applied for rapidly detecting various prohibited doping substances. Frequent modification is required as additional substances are prohibited. We developed and validated a non-target screening method requiring no further modification because it analyzes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service