Skip to Content
Merck

187712

Sigma-Aldrich

1-Chloroanthracene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9Cl
CAS Number:
Molecular Weight:
212.67
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

77-80 °C (lit.)

density

1.171 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

Clc1cccc2cc3ccccc3cc12

InChI

1S/C14H9Cl/c15-14-7-3-6-12-8-10-4-1-2-5-11(10)9-13(12)14/h1-9H

InChI key

SRIHSAFSOOUEGL-UHFFFAOYSA-N

General description

1-Chloroanthracene forms photodimer trans-bi(1-chloro-9,10-dihydro-9,10-anthracenediyl) via the solid-state [4+4]-photodimerization[1].

Application

1-Chloroanthracene was used to investigate the effect of polycyclic aromatic hydrocarbons on gap junctional intercellular communication and activation of intracellular receptor kinase in F344 rat liver epithelial cells[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Rummel et al.
Toxicological sciences : an official journal of the Society of Toxicology, 49(2), 232-240 (1999-07-23)
Many polycyclic aromatic hydrocarbons (PAHs) are known carcinogens. A considerable amount of research has been devoted to predicting the genotoxic, tumor-initiating potential of PAHs based on chemical structure. However, information on the correlation of structure with the non-genetoxic, epigenetic events
Ilona Turowska-Tyrk et al.
Acta crystallographica. Section C, Crystal structure communications, 60(Pt 2), o146-o148 (2004-02-10)
Crystals of the 1-chloroanthracene photodimer, viz. trans-bi(1-chloro-9,10-dihydro-9,10-anthracenediyl), C(28)H(18)Cl(2), were obtained from the solid-state [4+4]-photodimerization of the monomer, C(14)H(9)Cl, followed by recrystallization. The symmetry of the product molecules is defined by the orientation of the reactant molecules in the crystal. The
Miki Sato et al.
The Analyst, 142(23), 4560-4569 (2017-11-09)
The detection limits and photoionization thresholds of polycyclic aromatic hydrocarbons and their chlorides and nitrides on the water surface are examined using laser two-photon ionization and single-photon ionization, respectively. The laser two-photon ionization methods are highly surface-selective, with a high
Diógenes Herreno-Sáenz et al.
International journal of environmental research and public health, 3(2), 191-195 (2006-07-11)
Since the finding in the 1930s, a large number of polycyclic aromatic hydrocarbons (PAHs) of different structures have been tested for potential tumorigenicity. Structure-activity relationships of halo-PAHs have been investigated to determine the regions of a PAH that may be
Rong Jin et al.
Journal of chromatography. A, 1509, 114-122 (2017-06-18)
Isotopic dilution gas chromatography combined with high resolution mass spectrometry (GC/HRMS) has overwhelming advantages with respect to the accuracy of congener-specific ultratrace analysis of complex persistent organic pollutants (POPs) in environmental matrices. However, an isotopic dilution GC/HRMS method for analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service