Skip to Content
Merck
All Photos(2)

Key Documents

186023

Sigma-Aldrich

4-Chloro-7-(trifluoromethyl)quinoline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5ClF3N
CAS Number:
Molecular Weight:
231.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

69-71 °C (lit.)

solubility

chloroform: soluble 25 mg/mL, clear, colorless

functional group

chloro
fluoro

SMILES string

FC(F)(F)c1ccc2c(Cl)ccnc2c1

InChI

1S/C10H5ClF3N/c11-8-3-4-15-9-5-6(10(12,13)14)1-2-7(8)9/h1-5H

InChI key

LLRQVSZVVAKRJA-UHFFFAOYSA-N

Application

4-Chloro-7-(trifluoromethyl)quinolone was used in the synthesis of (4-substituted phenyl-1-piperazinyl) alkyl 2-aminobenzoates and 2-aminonicotinates[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P M Manoury et al.
Journal of medicinal chemistry, 22(5), 554-559 (1979-05-01)
A series of (4-substituted phenyl-1-piperazinyl)alkyl 2-aminobenzoates and 2-aminonicotinates has been prepared and screened for analgesic and antiinflammatory properties in mice and rats. The tabulated results reveal several 2-(4-substituted phenyl-1-piperazinyl)ethyl 2-(7- or 8-substituted 4-quinolinylamino)benzoates to be six to nine times more
Mostafa M Ghorab et al.
Acta pharmaceutica (Zagreb, Croatia), 64(3), 285-297 (2014-10-10)
Novel nineteen compounds based on a 4-aminoquinoline scaffold were designed and synthesized as potential antiproliferative agents. The new compounds were N-substituted at the 4-position by aryl or heteroaryl (1-9), quinolin- 3-yl (10), 2-methylquinolin-3-yl (11), thiazol-2-yl (12), and dapsone moieties (13

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service