Skip to Content
Merck
All Photos(1)

Key Documents

183679

Sigma-Aldrich

Methyl 1-benzyl-5-oxo-3-pyrrolidinecarboxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15NO3
CAS Number:
Molecular Weight:
233.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

97%

form

solid

mp

63-66 °C (lit.)

SMILES string

COC(=O)C1CN(Cc2ccccc2)C(=O)C1

InChI

1S/C13H15NO3/c1-17-13(16)11-7-12(15)14(9-11)8-10-5-3-2-4-6-10/h2-6,11H,7-9H2,1H3

InChI key

WTRWSSDZHQOPJI-UHFFFAOYSA-N

General description

Enantiomeric separation of methyl 1-benzyl-5-oxo-3-pyrrolidinecarboxylate has been achieved using methylchlorophenyl carbamate of cellulose and of amylose as polysaccharide chiral stationary phases by HPLC.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Review of chiral stationary phase development and chiral applications.
Beesley TE.
LC-GC, 24(5), 270-276 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service