Skip to Content
Merck
All Photos(1)

Key Documents

182745

Sigma-Aldrich

Poly(vinyl methyl ketone)

average Mw ~500,000 by GPC

Synonym(s):

3-Buten-2-one polymer, Atactic poly(methyl vinyl ketone), Methyl vinyl ketone polymer, Polybutenone

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€288.00

€288.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€288.00

About This Item

Linear Formula:
[CH2CH(COCH3)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€288.00


Please contact Customer Service for Availability

Request a Bulk Order

mol wt

average Mw ~500,000 by GPC

refractive index

n20/D 1.5 (lit.)

transition temp

Tg 28 °C

density

1.12 g/mL at 25 °C (lit.)

SMILES string

CC(=O)C=C

InChI

1S/C4H6O/c1-3-4(2)5/h3H,1H2,2H3

InChI key

FUSUHKVFWTUUBE-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service