Skip to Content
Merck
All Photos(1)

Key Documents

18241

Sigma-Aldrich

(R)-(−)-2-Amino-3-methylbutane

≥97.0% (GC)

Synonym(s):

(R)-(−)-3-Methyl-2-butylamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€75.60
5 G
€303.00

€75.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€75.60
5 G
€303.00

About This Item

Empirical Formula (Hill Notation):
C5H13N
CAS Number:
Molecular Weight:
87.16
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

€75.60


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)

optical activity

[α]/D −5.5±1°, neat

density

0.75 g/mL at 20 °C (lit.)

functional group

amine

SMILES string

CC(C)[C@@H](C)N

InChI

1S/C5H13N/c1-4(2)5(3)6/h4-5H,6H2,1-3H3/t5-/m1/s1

InChI key

JOZZAIIGWFLONA-RXMQYKEDSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

<69.8 °F

Flash Point(C)

< 21 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service