Skip to Content
Merck
All Photos(1)

Documents

180645

Sigma-Aldrich

2-Methoxyphenylacetonitrile

98%

Synonym(s):

2-Methoxybenzyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2CN
CAS Number:
Molecular Weight:
147.17
Beilstein:
2087748
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

crystals

bp

143 °C/15 mmHg (lit.)

mp

65-67 °C (lit.)

SMILES string

COc1ccccc1CC#N

InChI

1S/C9H9NO/c1-11-9-5-3-2-4-8(9)6-7-10/h2-5H,6H2,1H3

InChI key

DWJKILXTMUGXOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methoxyphenylacetonitrile was used in the synthesis of 2-(1-cyano-1-(2-methoxy)phenyl)methylidene-3-phenylthiazolidine-4,5-dione.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

264.2 °F - DIN 51758

Flash Point(C)

129 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uwe Albrecht et al.
Bioorganic & medicinal chemistry, 13(14), 4402-4407 (2005-05-21)
2-Alkylidenethiazolidine-4,5-diones were prepared by novel one-pot cyclizations of arylacetonitriles with isothiocyanates and ethyl 2-chloro-2-oxoacetate. The products show antibiotic activity against the Gram-positive bacteria Bacillus subtilis and Staphylococcus aureus.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service