Skip to Content
Merck
All Photos(1)

Key Documents

180335

Supelco

3-(Trimethylsilyl)propionic acid sodium salt

99%

Synonym(s):

Sodium 3-(trimethylsilyl)propionate, TMSP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCH2CH2CO2Na
CAS Number:
Molecular Weight:
168.24
Beilstein:
4021529
EC Number:
MDL number:
UNSPSC Code:
12000000
Pricing and availability is not currently available.

Assay

99%

SMILES string

[Na+].C[Si](C)(C)CC[13C]([O-])=O

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service