Skip to Content
Merck
All Photos(2)

Key Documents

177547

Sigma-Aldrich

2,5-Dichlorobenzenesulfonyl chloride

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€108.00
100 G
€411.00

€108.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€108.00
100 G
€411.00

About This Item

Linear Formula:
Cl2C6H3SO2Cl
CAS Number:
Molecular Weight:
245.51
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€108.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

mp

36-37 °C (lit.)

functional group

chloro

SMILES string

Clc1ccc(Cl)c(c1)S(Cl)(=O)=O

InChI

1S/C6H3Cl3O2S/c7-4-1-2-5(8)6(3-4)12(9,10)11/h1-3H

InChI key

BXCOSWRSIISQSL-UHFFFAOYSA-N

Application

2,5-Dichlorobenzenesulfonyl chloride was used to study the synthesis and structure–activity profile of benzoxazole benzenesulfonamide analogs[1].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunqiu Lai et al.
Bioorganic & medicinal chemistry letters, 16(7), 1807-1810 (2006-02-01)
A series of novel benzoxazole benzenesulfonamides was synthesized as inhibitors of fructose-1,6-bisphosphatase (FBPase-1). Extensive SAR studies led to a potent inhibitor, 53, with an IC(50) of 0.57microM. Compound 17 exhibited excellent bioavailability and a good pharmacokinetic profile in rats.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service