Skip to Content
Merck
All Photos(1)

Key Documents

176745

Sigma-Aldrich

Perchloric acid-d solution

68 wt. % in D2O, 99 atom % D

Synonym(s):

Deuteroperchloric acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€133.00
25 G
€1,520.00

€133.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 G
€133.00
25 G
€1,520.00

About This Item

Linear Formula:
DClO4 / D2O
CAS Number:
Molecular Weight:
101.46
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.12

€133.00


Please contact Customer Service for Availability

Request more information

isotopic purity

99 atom % D

Quality Level

reaction suitability

reagent type: oxidant

concentration

68 wt. % in D2O

technique(s)

NMR: suitable

refractive index

n20/D 1.4165

density

1.680 g/mL at 25 °C

mass shift

M+1

SMILES string

[2H]OCl(=O)(=O)=O

InChI

1S/ClHO4/c2-1(3,4)5/h(H,2,3,4,5)/i/hD

InChI key

VLTRZXGMWDSKGL-DYCDLGHISA-N

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Liq. 1 - Skin Corr. 1A

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service